2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile 97%

Lieferant: Apollo Scientific

PC302052-1G PC302052-25G
APOSPC302052-1GEA 21.9 EUR
APOSPC302052-1G APOSPC302052-25G
2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile 97%
2-(3-Chloro-5-(trifluoromethyl)phenyl)acetonitrile
Formel: C₉H₅ClF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD04115850
CAS-Nummer: 886496-95-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR