3-Fluoro-4-methoxycinnamic acid 97%

Lieferant: Apollo Scientific
Achtung

PC302055-1G
APOSPC302055-1GEA 0 EUR
APOSPC302055-1G
3-Fluoro-4-methoxycinnamic acid 97%
3-Fluoro-4-methoxycinnamic acid
Formel: C₁₀H₉FO₃
Schmelzpunkt: 227…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03002809
CAS-Nummer: 713-85-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR