5-Chloro-2-(trifluoromethyl)cinnamic acid 97%

Lieferant: Apollo Scientific

PC302066-5G PC302066-1G
APOSPC302066-5GEA 0 EUR
APOSPC302066-5G APOSPC302066-1G
5-Chloro-2-(trifluoromethyl)cinnamic acid 97%
5-Chloro-2-(trifluoromethyl)cinnamic acid
Formel: C₁₀H₆ClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06205353
CAS-Nummer: 773129-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR