2-Chlor-3,6-difluorbenzonitril 98%

Lieferant: Apollo Scientific
Achtung

PC302072-25G PC302072-5G PC302072-1G
APOSPC302072-25GEA 264 EUR
APOSPC302072-25G APOSPC302072-5G APOSPC302072-1G
2-Chlor-3,6-difluorbenzonitril 98%
2-Chlor-3,6-difluorbenzonitril
Formel: C₇H₂ClF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD06660175
CAS-Nummer: 886501-33-3
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR