2-Chlor-3,6-difluorbenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302072-25G
PC302072-5G
PC302072-1G
APOSPC302072-25GEA
264
EUR
APOSPC302072-25G
APOSPC302072-5G
APOSPC302072-1G
2-Chlor-3,6-difluorbenzonitril 98%
2-Chlor-3,6-difluorbenzonitril
Formel:
C₇H₂ClF₂N Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660175 CAS-Nummer: 886501-33-3 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...