2-Fluoro-5-methylphenylacetonitrile 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Fluoro-5-methylbenzyl cyanide, (2-Fluoro-5-methylphenyl)acetonitrile

PC302077-1G
APOSPC302077-1GEA 299 EUR
APOSPC302077-1G
2-Fluoro-5-methylphenylacetonitrile 97%
2-Fluoro-5-methylphenylacetonitrile
Formel: C₉H₈FN
Molecular Weight: 149,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772783
CAS-Nummer: 945541-97-9
UN: 3276
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR