2-Methoxy-3-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific

PC302078-1G PC302078-5G PC302078-25G PC302078-100G
APOSPC302078-1GEA 0 EUR
APOSPC302078-1G APOSPC302078-5G APOSPC302078-25G APOSPC302078-100G
2-Methoxy-3-(trifluormethyl)benzylbromid 98%
2-Methoxy-3-(trifluormethyl)benzylbromid
Formel: C₉H₈BrF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09832315
CAS-Nummer: 1017778-77-6
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR