2-Methoxy-6-(trifluormethyl)benzylalkohol 98%

Lieferant: Apollo Scientific

PC302079-1G PC302079-5G PC302079-10G
APOSPC302079-1GEA 126 EUR
APOSPC302079-1G APOSPC302079-5G APOSPC302079-10G
2-Methoxy-6-(trifluormethyl)benzylalkohol 98%
2-Methoxy-6-(trifluormethyl)benzylalkohol
Formel: C₉H₉F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832325
CAS-Nummer: 1017779-01-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR