2-Methyl-3-(trifluormethyl)zimtsäure 98%

Lieferant: Apollo Scientific

PC302080-1G PC302080-5G PC302080-10G
APOSPC302080-1GEA 87 EUR
APOSPC302080-1G APOSPC302080-5G APOSPC302080-10G
2-Methyl-3-(trifluormethyl)zimtsäure 98%
2-Methyl-3-(trifluormethyl)zimtsäure
Formel: C₁₁H₉F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832281
CAS-Nummer: 1017777-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR