3,5-Difluor-2-methoxybenzylamin 98%

Lieferant: Apollo Scientific

PC302084-1G PC302084-5G PC302084-10G
APOSPC302084-1GEA 0 EUR
APOSPC302084-1G APOSPC302084-5G APOSPC302084-10G
3,5-Difluor-2-methoxybenzylamin 98%
3,5-Difluor-2-methoxybenzylamin
Formel: C₈H₉F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06660178
CAS-Nummer: 886502-00-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR