3-Methoxy-2-(trifluormethyl)benzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302096-1G
PC302096-5G
PC302096-10G
APOSPC302096-1GEA
0
EUR
APOSPC302096-1G
APOSPC302096-5G
APOSPC302096-10G
3-Methoxy-2-(trifluormethyl)benzonitril 98%
3-Methoxy-2-(trifluormethyl)benzonitril
Formel:
C₉H₆F₃NO Schmelzpunkt: 103…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185684 CAS-Nummer: 1214385-02-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...