4-n-Butoxy-2,6-difluorobenzoyl chloride 97%

Lieferant: Apollo Scientific

PC302101-1G
APOSPC302101-1GEA 98.6 EUR
APOSPC302101-1G
4-n-Butoxy-2,6-difluorobenzoyl chloride 97%
4-n-Butoxy-2,6-difluorobenzoyl chloride
Formel: C₁₁H₁ClF₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD22201027
CAS-Nummer: 1373920-74-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR