4-Fluoro-3-methyl-5-(trifluoromethyl)cinnamic acid 97%

Lieferant: Apollo Scientific

Synonyme: 4-Fluoro-3-methyl-5-(trifluoromethyl)cinnamic acid

PC302113-1G
APOSPC302113-1GEA 98.6 EUR
APOSPC302113-1G
4-Fluoro-3-methyl-5-(trifluoromethyl)cinnamic acid 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)cinnamic acid
Formel: C₁₁H₈F₄O₂
Molecular Weight: 248.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201075
CAS-Nummer: 1373921-81-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR