4'-Fluor-3'-methylacetophenon 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Fluoro-3-methylphenyl)ethanone

PC302115-25G PC302115-5G PC302115-100G
APOSPC302115-25GEA 119 EUR
APOSPC302115-25G APOSPC302115-5G APOSPC302115-100G
4'-Fluor-3'-methylacetophenon 95%
4'-Fluor-3'-methylacetophenon
Formel: C₉H₉FO
Lagertemperatur: Raumtemperatur
MDL: MFCD04972879
CAS-Nummer: 369-32-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR