2,3-Difluoro-4-methyl-DL-phenylalanine

Lieferant: Apollo Scientific

PC302121-1G PC302121-5G PC302121-10G
APOSPC302121-1GEA 163 EUR
APOSPC302121-1G APOSPC302121-5G APOSPC302121-10G
2,3-Difluoro-4-methyl-DL-phenylalanine
2,3-Difluoro-4-methyl-DL-phenylalanine
Formel: C₁₀H₁₁F₂NO₂
Molecular Weight: 215.197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660244
CAS-Nummer: 1215491-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR