2-Chloro-4-fluoro-DL-phenylalanine

Lieferant: Apollo Scientific

PC302127-1G PC302127-5G PC302127-10G
APOSPC302127-1GEA 116 EUR
APOSPC302127-1G APOSPC302127-5G APOSPC302127-10G
2-Chloro-4-fluoro-DL-phenylalanine
2-Chloro-4-fluoro-DL-phenylalanine
Formel: C₉H₉ClFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06660324
CAS-Nummer: 754152-25-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR