3-Chloro-2,6-difluoro-DL-phenylalanine
Lieferant: Apollo Scientific
PC302138-1G
PC302138-5G
PC302138-10G
APOSPC302138-1GEA
116
EUR
APOSPC302138-1G
APOSPC302138-5G
APOSPC302138-10G
3-Chloro-2,6-difluoro-DL-phenylalanine
3-Chloro-2,6-difluoro-DL-phenylalanine
Formel:
C₉H₈ClF₂NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116006 CAS-Nummer: 1260006-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...