3-Chloro-2,6-difluoro-DL-phenylalanine

Lieferant: Apollo Scientific

PC302138-1G PC302138-5G PC302138-10G
APOSPC302138-1GEA 116 EUR
APOSPC302138-1G APOSPC302138-5G APOSPC302138-10G
3-Chloro-2,6-difluoro-DL-phenylalanine
3-Chloro-2,6-difluoro-DL-phenylalanine
Formel: C₉H₈ClF₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04116006
CAS-Nummer: 1260006-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR