3-Fluoro-2-methyl-DL-phenylalanine

Lieferant: Apollo Scientific

PC302140-1G PC302140-5G
APOSPC302140-1GEA 116 EUR
APOSPC302140-1G APOSPC302140-5G
3-Fluoro-2-methyl-DL-phenylalanine
3-Fluoro-2-methyl-DL-phenylalanine
Formel: C₁₀H₁₂FNO₂
Molecular Weight: 197.206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660341
CAS-Nummer: 1260004-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR