3-Fluoro-4-(trifluoromethyl)-DL-phenylalanine

Lieferant: Apollo Scientific
Achtung

PC302141-1G PC302141-5G PC302141-10G
APOSPC302141-1GEA 116 EUR
APOSPC302141-1G APOSPC302141-5G APOSPC302141-10G
3-Fluoro-4-(trifluoromethyl)-DL-phenylalanine
3-Fluoro-4-(trifluoromethyl)-DL-phenylalanine
Formel: C₁₀H₉F₄NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04115998
CAS-Nummer: 1215491-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR