3-Fluoro-5-methyl-DL-phenylalanine

Lieferant: Apollo Scientific

PC302144-1G PC302144-5G
APOSPC302144-1GEA 128 EUR
APOSPC302144-1G APOSPC302144-5G
3-Fluoro-5-methyl-DL-phenylalanine
3-Fluoro-5-methyl-DL-phenylalanine
Formel: C₁₀H₁₂FNO₂
Molecular Weight: 197.206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05227967
CAS-Nummer: 603106-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR