4-Fluoro-3-methyl-DL-phenylalanine
Lieferant: Apollo Scientific
|
Achtung
|
PC302151-1G
PC302151-5G
PC302151-10G
PC302151-25G
PC302151-100G
APOSPC302151-1GEA
128
EUR
APOSPC302151-1G
APOSPC302151-5G
APOSPC302151-10G
APOSPC302151-25G
APOSPC302151-100G
4-Fluoro-3-methyl-DL-phenylalanine
4-Fluoro-3-methyl-DL-phenylalanine
Formel:
C₁₀H₁₂FNO₂ Molecular Weight: 197.206 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116031 CAS-Nummer: 1259956-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...