2,4-Dibrom-5-(trifluormethoxy)anisol 97%
Lieferant: Apollo Scientific
PC302156-25G
PC302156-5G
PC302156-1G
APOSPC302156-25GEA
360
EUR
APOSPC302156-25G
APOSPC302156-5G
APOSPC302156-1G
2,4-Dibrom-5-(trifluormethoxy)anisol 97%
2,4-Dibrom-5-(trifluormethoxy)anisol
Formel:
C₈H₅Br₂F₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22201036 CAS-Nummer: 1373920-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...