2-Brom-4-chlor-6-fluorphenol 98%

Lieferant: Apollo Scientific

PC302165-5G
APOSPC302165-5GEA 36.4 EUR
APOSPC302165-5G
2-Brom-4-chlor-6-fluorphenol 98%
2-Brom-4-chlor-6-fluorphenol
Formel: C₆H₃BrClFO
Molecular Weight: 225,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115955
CAS-Nummer: 886499-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR