2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetonitrile 97%

Lieferant: Apollo Scientific

PC302172-5G
APOSPC302172-5GEA 306 EUR
APOSPC302172-5G
2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetonitrile 97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetonitrile
Formel: C₁₀H₇F₄NO
Lagertemperatur: Raumtemperatur
MDL: MFCD22201045
CAS-Nummer: 1373920-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR