2-Methoxy-4-(trifluoromethoxy)benzaldehyde 97%

Lieferant: Apollo Scientific
Achtung

PC302173-10G PC302173-5G PC302173-25G
APOSPC302173-10GEA 0 EUR
APOSPC302173-10G APOSPC302173-5G APOSPC302173-25G
2-Methoxy-4-(trifluoromethoxy)benzaldehyde 97%
2-Methoxy-4-(trifluoromethoxy)benzaldehyde
Formel: C₉H₇F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04115965
CAS-Nummer: 886500-13-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR