2-Methoxy-4-(trifluormethoxy)phenylessigsäure 97%

Lieferant: Apollo Scientific

PC302175-1G
APOSPC302175-1GEA 95.7 EUR
APOSPC302175-1G
2-Methoxy-4-(trifluormethoxy)phenylessigsäure 97%
2-Methoxy-4-(trifluormethoxy)phenylessigsäure
Formel: C₁₀H₉F₃O₄
Molecular Weight: 250.17 g/mol
Schmelzpunkt: 103…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115972
CAS-Nummer: 886500-45-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR