2-Methoxy-5-(trifluormethoxy)benzonitril 97%

Lieferant: Apollo Scientific

PC302176-5G PC302176-1G PC302176-25G
APOSPC302176-5GEA 0 EUR
APOSPC302176-5G APOSPC302176-1G APOSPC302176-25G
2-Methoxy-5-(trifluormethoxy)benzonitril 97%
2-Methoxy-5-(trifluormethoxy)benzonitril
Formel: C₉H₆F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04115963
CAS-Nummer: 886500-03-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR