3,5-Difluoro-4-methoxycinnamic acid 98%

Lieferant: Apollo Scientific

PC302185-1G
APOSPC302185-1GEA 126 EUR
APOSPC302185-1G
3,5-Difluoro-4-methoxycinnamic acid 98%
3,5-Difluoro-4-methoxycinnamic acid
Formel: C₁₀H₈F₂O₃
Molecular Weight: 214,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115917
CAS-Nummer: 105219-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR