3-[2-Methyl-4-(trifluoromethoxy)phenyl]propionic acid

Lieferant: Apollo Scientific

PC302188-1G
APOSPC302188-1GEA 0 EUR
APOSPC302188-1G
3-[2-Methyl-4-(trifluoromethoxy)phenyl]propionic acid
3-[2-Methyl-4-(trifluoromethoxy)phenyl]propionic acid
Formel: C₁₁H₁₁F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22201060
CAS-Nummer: 1373921-13-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR