3-[2-Methyl-5-(trifluoromethoxy)phenyl]propionic acid 97%

Lieferant: Apollo Scientific

Synonyme: Benzenepropanoic acid, 2-methyl-4-(trifluoromethoxy)-, 3-[2-Methyl-4-(trifluoromethoxy)phenyl]propanoic acid

PC302189-1G
APOSPC302189-1GEA 112 EUR
APOSPC302189-1G
3-[2-Methyl-5-(trifluoromethoxy)phenyl]propionic acid 97%
3-[2-Methyl-5-(trifluoromethoxy)phenyl]propionic acid
Formel: C₁₁H₁₁F₃O₃
Molecular Weight: 248,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201057
CAS-Nummer: 1373921-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR