(3-Chlor-5-(trifluormethyl)phenyl)essigsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2-(3-Chloro-5-(trifluoromethyl)phenyl)acetic acid

PC302198-1G PC302198-5G PC302198-10G
APOSPC302198-1GEA 37.7 EUR
APOSPC302198-1G APOSPC302198-5G APOSPC302198-10G
(3-Chlor-5-(trifluormethyl)phenyl)essigsäure 98%
(3-Chlor-5-(trifluormethyl)phenyl)essigsäure
Formel: C₉H₆ClF₃O₂
Molecular Weight: 238.59 g/mol
Schmelzpunkt: 125…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115851
CAS-Nummer: 886496-99-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR