4,6-Difluoroindole-2-carboxylic acid 97%
Lieferant: Apollo Scientific
Synonyme:
4,6-Difluoro-1H-indole-2-carboxylic acid
PC3022-5G
APOSPC3022-5GEA
95.7
EUR
APOSPC3022-5G
4,6-Difluoroindole-2-carboxylic acid 97%
4,6-Difluoroindole-2-carboxylic acid
Formel:
C₉H₅F₂NO₂ Molecular Weight: 197,1400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863158 CAS-Nummer: 247564-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...