3-Fluoro-5-(pentafluorothio)benzoyl chloride 97%
Lieferant: Apollo Scientific
PC302201-1G
APOSPC302201-1GEA
0
EUR
APOSPC302201-1G
3-Fluoro-5-(pentafluorothio)benzoyl chloride 97%
3-Fluoro-5-(pentafluorothio)benzoyl chloride
Formel:
C₇H₃ClF₆OS Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652427 CAS-Nummer: 1240256-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...