3-Fluoro-5-(trifluoromethoxy)cinnamic acid 98%

Lieferant: Apollo Scientific

PC302202-1G
APOSPC302202-1GEA 114 EUR
APOSPC302202-1G
3-Fluoro-5-(trifluoromethoxy)cinnamic acid 98%
3-Fluoro-5-(trifluoromethoxy)cinnamic acid
Formel: C₁₀H₆F₄O₃
Molecular Weight: 250,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18783232
CAS-Nummer: 1353001-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR