4-Ethoxy-3-(trifluoromethyl)benzamide 98%

Lieferant: Apollo Scientific

PC302210-1G
APOSPC302210-1GEA 29 EUR
APOSPC302210-1G
4-Ethoxy-3-(trifluoromethyl)benzamide 98%
4-Ethoxy-3-(trifluoromethyl)benzamide
Formel: C₁₀H₁₀F₃NO₂
Molecular Weight: 233,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13196583
CAS-Nummer: 1206593-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR