4-Ethoxy-3,5-difluorocinnamic acid 97%

Lieferant: Apollo Scientific

PC302211-1G
APOSPC302211-1GEA 105 EUR
APOSPC302211-1G
4-Ethoxy-3,5-difluorocinnamic acid 97%
4-Ethoxy-3,5-difluorocinnamic acid
Formel: C₁₁H₁₀F₂O₃
Molecular Weight: 228,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258693
CAS-Nummer: 1017779-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR