4-Ethoxy-3,5-difluorphenol 97%

Lieferant: Apollo Scientific

PC302212-1G PC302212-5G
APOSPC302212-1GEA 105 EUR
APOSPC302212-1G APOSPC302212-5G
4-Ethoxy-3,5-difluorphenol 97%
4-Ethoxy-3,5-difluorphenol
Formel: C₈H₈F₂O₂
Molecular Weight: 174.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258694
CAS-Nummer: 1017779-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR