4-Fluoro-3-(trifluoromethoxy)cinnamic acid 98%

Lieferant: Apollo Scientific

PC302213-1G PC302213-5G
APOSPC302213-1GEA 148 EUR
APOSPC302213-1G APOSPC302213-5G
4-Fluoro-3-(trifluoromethoxy)cinnamic acid 98%
4-Fluoro-3-(trifluoromethoxy)cinnamic acid
Formel: C₁₀H₆F₄O₃
Molecular Weight: 250.146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660259
CAS-Nummer: 886501-38-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR