4-Fluoro-3,5-dimethylcinnamic acid 97%

Lieferant: Apollo Scientific

PC302215-1G
APOSPC302215-1GEA 574 EUR
APOSPC302215-1G
4-Fluoro-3,5-dimethylcinnamic acid 97%
4-Fluoro-3,5-dimethylcinnamic acid
Formel: C₁₁H₁₁FO₂
Molecular Weight: 194.202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832376
CAS-Nummer: 1429551-15-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR