4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetonitrile

PC302217-1G
APOSPC302217-1GEA 112 EUR
APOSPC302217-1G
4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetonitrile 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetonitrile
Formel: C₁₀H₇F₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD22201073
CAS-Nummer: 1373920-75-2
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR