2,4-Bis(trifluormethyl)zimtsäure 98%

Lieferant: Apollo Scientific

PC302223-1G PC302223-5G PC302223-10G
APOSPC302223-1GEA 219 EUR
APOSPC302223-1G APOSPC302223-5G APOSPC302223-10G
2,4-Bis(trifluormethyl)zimtsäure 98%
2,4-Bis(trifluormethyl)zimtsäure
Formel: C₁₁H₆F₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06205287
CAS-Nummer: 773129-10-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR