2-Chloro-5-(difluoromethoxy)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC302227-1G PC302227-5G PC302227-10G
APOSPC302227-1GEA 21.9 EUR
APOSPC302227-1G APOSPC302227-5G APOSPC302227-10G
2-Chloro-5-(difluoromethoxy)benzyl bromide 95%
2-Chloro-5-(difluoromethoxy)benzyl bromide
Formel: C₈H₆BrClF₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09832368
CAS-Nummer: 1017779-54-2
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR