2-Chloro-6-(trifluoromethoxy)aniline 98%

Lieferant: Apollo Scientific
Achtung

PC302230-1G PC302230-5G PC302230-10G PC302230-25G PC302230-100G
APOSPC302230-1GEA 92.8 EUR
APOSPC302230-1G APOSPC302230-5G APOSPC302230-10G APOSPC302230-25G APOSPC302230-100G
2-Chloro-6-(trifluoromethoxy)aniline 98%
2-Chloro-6-(trifluoromethoxy)aniline
Formel: C₇H₅ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD07779302
CAS-Nummer: 1261673-70-4
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR