2-Fluor-4-(pentafluorthio)benzoylchlorid 97%
Lieferant: Apollo Scientific
PC302237-1G
APOSPC302237-1GEA
348
EUR
APOSPC302237-1G
2-Fluor-4-(pentafluorthio)benzoylchlorid 97%
2-Fluor-4-(pentafluorthio)benzoylchlorid
Formel:
C₇H₃ClF₆OS Molecular Weight: 284,60 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652505 CAS-Nummer: 1240256-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...