2-Fluoro-4-(pentafluorothio)benzyl alcohol

Lieferant: Apollo Scientific

PC302238-1G
APOSPC302238-1GEA 0 EUR
APOSPC302238-1G
2-Fluoro-4-(pentafluorothio)benzyl alcohol
2-Fluoro-4-(pentafluorothio)benzyl alcohol
Formel: C₇H₆F₆OS
Lagertemperatur: Raumtemperatur
MDL: MFCD16652506
CAS-Nummer: 1240257-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR