2-Fluor-4-(pentafluorthio)benzylamin 98%

Lieferant: Apollo Scientific

PC302239-1G
APOSPC302239-1GEA 0 EUR
APOSPC302239-1G
2-Fluor-4-(pentafluorthio)benzylamin 98%
2-Fluor-4-(pentafluorthio)benzylamin
Formel: C₇H₇F₆NS
Lagertemperatur: Raumtemperatur
MDL: MFCD16652513
CAS-Nummer: 1240257-86-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR