2-Fluor-5-methyl-4-(trifluormethyl)zimtsäure 98%

Lieferant: Apollo Scientific

PC302245-1G
APOSPC302245-1GEA 0 EUR
APOSPC302245-1G
2-Fluor-5-methyl-4-(trifluormethyl)zimtsäure 98%
2-Fluor-5-methyl-4-(trifluormethyl)zimtsäure
Formel: C₁₁H₈F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19687201
CAS-Nummer: 1324063-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR