2-Methoxy-3-(trifluormethyl)zimtsäure 97%

Lieferant: Apollo Scientific

PC302247-1G
APOSPC302247-1GEA 0 EUR
APOSPC302247-1G
2-Methoxy-3-(trifluormethyl)zimtsäure 97%
2-Methoxy-3-(trifluormethyl)zimtsäure
Formel: C₁₁H₉F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11519316
CAS-Nummer: 1092460-72-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR