2-Methoxy-3-(trifluormethyl)phenylacetonitril 97%

Lieferant: Apollo Scientific
Achtung

PC302248-1G PC302248-5G PC302248-25G
APOSPC302248-1GEA 0 EUR
APOSPC302248-1G APOSPC302248-5G APOSPC302248-25G
2-Methoxy-3-(trifluormethyl)phenylacetonitril 97%
2-Methoxy-3-(trifluormethyl)phenylacetonitril
Formel: C₁₀H₈F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09832316
CAS-Nummer: 1017778-81-2
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR